Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B3gat1
Entrez Gene ID
117108
Gene Name
beta-1,3-glucuronyltransferase 1
Gene Aliases
Hnk-1
Chromosome Location
Chr.8: 27776955 - 27804368 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054003.1 NP_446455.1 5 - 6 1155 58
XM_006242733.2 XP_006242795.1 5 - 6 1427 58
XM_006242734.2 XP_006242796.1 5 - 6 1416 58
GenBank mRNA D88035.1 - 5 - 6 1155 58

View Full Product Details