The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133405.2 | NP_596896.2 | 10 - 11 | 1637 | 54 |
GenBank mRNA | AY033877.1 | - | 10 - 11 | 1647 | 54 |
BC166416.1 | - | 10 - 11 | 1637 | 54 | |
FQ211347.1 | - | 10 - 11 | 1647 | 54 |