Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cry2
Entrez Gene ID
170917
Gene Name
cryptochrome circadian clock 2
Gene Aliases
-
Chromosome Location
Chr.3: 81314151 - 81344109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133405.2 NP_596896.2 2 - 3 323 109
GenBank mRNA AY033877.1 - 2 - 3 333 109
BC166416.1 - 2 - 3 323 109
FQ211347.1 - 2 - 3 333 109

View Full Product Details