Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ehd4
Entrez Gene ID
192204
Gene Name
EH-domain containing 4
Gene Aliases
Past2
Chromosome Location
Chr.3: 111877853 - 111941841 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139324.1 NP_647540.1 2 - 3 427 75
GenBank mRNA AY094093.1 - 2 - 3 427 75
BC083175.1 - 2 - 3 409 75

View Full Product Details