Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lamp1
Entrez Gene ID
25328
Gene Name
lysosomal-associated membrane protein 1
Gene Aliases
LGP120
Chromosome Location
Chr.16: 81689576 - 81706561 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012857.2 NP_036989.1 8 - 9 1191 75
GenBank mRNA M34959.1 - 8 - 9 1191 75
X14765.1 - 7 - 8 1027 75

View Full Product Details