Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Anxa5
Entrez Gene ID
25673
Gene Name
annexin A5
Gene Aliases
Anx5, LC5
Chromosome Location
Chr.2: 123162477 - 123193178 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013132.1 NP_037264.1 12 - 13 971 64
XM_006232268.2 XP_006232330.1 12 - 13 1106 64
GenBank mRNA BC081855.1 - 12 - 13 1024 64
FQ213209.1 - 13 - 14 1101 64
FQ214176.1 - 12 - 13 1003 64
FQ229127.1 - 12 - 13 959 64
M21730.1 - 12 - 13 971 64

View Full Product Details