The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017012.1 | NP_058708.1 | 8 - 9 | 2890 | 77 |
GenBank mRNA | D10891.1 | - | 8 - 9 | 2890 | 77 |