Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pccb
Entrez Gene ID
24624
Gene Name
propionyl CoA carboxylase, beta polypeptide
Gene Aliases
-
Chromosome Location
Chr.8: 109368887 - 109418871 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017030.2 NP_058726.2 11 - 12 1213 78
GenBank mRNA BC078855.1 - 11 - 12 1213 78
M14634.1 - 11 - 12 1214 78

View Full Product Details