Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdlim4
Entrez Gene ID
24915
Gene Name
PDZ and LIM domain 4
Gene Aliases
H-Rev18, Ril
Chromosome Location
Chr.10: 39390578 - 39405311 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017062.2 NP_058758.1 4 - 5 581 62
XM_006246230.2 XP_006246292.1 4 - 5 926 62
XM_006246231.2 XP_006246293.1 4 - 5 639 62
GenBank mRNA X76454.1 - 4 - 5 582 62

View Full Product Details