The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017105.1 | NP_058801.1 | 2 - 3 | 1817 | 60 |
GenBank mRNA | D63860.1 | - | 2 - 3 | 1817 | 60 |