Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc7a3
Entrez Gene ID
29485
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 3
Gene Aliases
Atrc3, Cat3, Slc7a2, cat-3
Chromosome Location
Chr.X: 70972767 - 70978952 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017217.1 NP_058913.1 9 - 10 1640 79
XM_006257059.2 XP_006257121.1 10 - 11 1970 79
GenBank mRNA AB000113.1 - 9 - 10 1640 79

View Full Product Details