Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pitpna
Entrez Gene ID
29525
Gene Name
phosphatidylinositol transfer protein, alpha
Gene Aliases
Pitpn
Chromosome Location
Chr.10: 63731767 - 63772049 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017231.1 NP_058927.1 8 - 9 732 73
GenBank mRNA BC070945.1 - 8 - 9 760 73
FQ235021.1 - 2 - 3 133 73
M25758.1 - 8 - 9 732 73

View Full Product Details