Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Car4
Entrez Gene ID
29242
Gene Name
carbonic anhydrase 4
Gene Aliases
Ca4
Chromosome Location
Chr.10: 72272286 - 72281069 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019174.3 NP_062047.1 4 - 5 525 77
GenBank mRNA BC097329.1 - 4 - 5 420 77
CB746957.1 - 1 - 2 86 77
FQ222059.1 - - 475 77
S68245.1 - 4 - 5 468 77

View Full Product Details