The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019269.1 | NP_062142.1 | 9 - 10 | 1685 | 68 |
GenBank mRNA | AB017260.1 | - | 9 - 10 | 1685 | 68 |
AF110416.1 | - | 9 - 10 | 1685 | 68 | |
AJ001933.1 | - | 9 - 10 | 1671 | 68 | |
BC166421.1 | - | 8 - 9 | 1434 | 68 |