Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Negr1
Entrez Gene ID
59318
Gene Name
neuronal growth regulator 1
Gene Aliases
-
Chromosome Location
Chr.2: 262914240 - 263650441 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021682.1 NP_067714.1 4 - 5 742 76
GenBank mRNA AB017139.1 - 4 - 5 742 76

View Full Product Details