Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fstl1
Entrez Gene ID
79210
Gene Name
follistatin-like 1
Gene Aliases
Frp, Fstl
Chromosome Location
Chr.11: 65792192 - 65845375 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024369.2 NP_077345.1 4 - 5 380 78
GenBank mRNA AY325203.1 - - 479 78
BC087014.1 - 4 - 5 380 78
U06864.1 - 4 - 5 355 78

View Full Product Details