Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nsfl1c
Entrez Gene ID
83809
Gene Name
NSFL1 (p97) cofactor (p47)
Gene Aliases
XY40, p47
Chromosome Location
Chr.3: 146980923 - 147005475 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031981.2 NP_114187.1 8 - 9 1055 61
XM_006235254.2 XP_006235316.1 9 - 10 1028 61
GenBank mRNA AB002086.1 - 8 - 9 1055 61
BC072464.1 - 8 - 9 952 61
Y10769.1 - 8 - 9 952 61

View Full Product Details