Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnd3
Entrez Gene ID
65195
Gene Name
potassium channel, voltage-gated Shal-related subfamily D, member 3
Gene Aliases
Kv4.3
Chromosome Location
Chr.2: 207923775 - 208140727 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270963.1 NP_001257892.1 5 - 6 1555 69
NM_031739.3 NP_113927.2 5 - 6 1555 69
GenBank mRNA L48619.1 - 5 - 6 1562 69
U75448.1 - 4 - 5 1483 69

View Full Product Details