The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177931.2 | NP_808792.1 | 2 - 3 | 222 | 130 |
XM_006238515.2 | XP_006238577.1 | 3 - 4 | 424 | 130 | |
XM_006238516.2 | XP_006238578.1 | 4 - 5 | 537 | 130 | |
XM_006238518.2 | XP_006238580.1 | 3 - 4 | 424 | 130 | |
GenBank mRNA | AB105378.1 | - | 2 - 3 | 222 | 130 |