The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181371.2 | NP_852036.1 | 4 - 5 | 420 | 83 |
XM_006236391.2 | XP_006236453.1 | 3 - 4 | 362 | 83 | |
GenBank mRNA | FJ179400.1 | - | 4 - 5 | 386 | 83 |
FQ211030.1 | - | 4 - 5 | 438 | 83 | |
FQ214179.1 | - | 4 - 5 | 503 | 83 | |
FQ220456.1 | - | 4 - 5 | 476 | 83 | |
S83436.1 | - | 4 - 5 | 420 | 83 |