The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181372.2 | NP_852037.1 | 1 - 2 | 201 | 127 |
XM_006237081.2 | XP_006237143.1 | 3 - 4 | 506 | 127 | |
XM_006237082.2 | XP_006237144.1 | 2 - 3 | 588 | 127 | |
XM_008763311.1 | XP_008761533.1 | 2 - 3 | 585 | 127 | |
GenBank mRNA | AF486185.1 | - | 1 - 2 | 142 | 127 |
AF486186.1 | - | 1 - 2 | 201 | 127 | |
AF486187.1 | - | 2 - 3 | 289 | 127 | |
FQ221012.1 | - | 2 - 3 | 299 | 127 |