The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012683.2 | NP_036815.1 | 1 - 2 | 887 | 68 |
GenBank mRNA | AF461736.1 | - | 1 - 2 | 890 | 68 |
AY435128.1 | - | 1 - 2 | 871 | 68 |