The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022952.2 | NP_075241.2 | 2 - 3 | 218 | 63 |
GenBank mRNA | BC088138.1 | - | 2 - 3 | 218 | 63 |
FQ234449.1 | - | 2 - 3 | 237 | 63 | |
M37194.1 | - | 2 - 3 | 220 | 63 | |
U75917.1 | - | 2 - 3 | 237 | 63 |