Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cxcl11
Entrez Gene ID
305236
Gene Name
chemokine (C-X-C motif) ligand 11
Gene Aliases
SCYB11
Chromosome Location
Chr.14: 17195395 - 17198170 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182952.2 NP_891997.1 3 - 4 317 75
GenBank mRNA AF537209.1 - 3 - 4 317 75
AY340181.1 - 3 - 4 296 75
BC168725.1 - 3 - 4 325 75

View Full Product Details