The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177305.1 | NP_001170776.1 | 3 - 4 | 384 | 81 |
NM_001271536.1 | NP_001258465.1 | 3 - 4 | 470 | 81 | |
NM_012495.2 | NP_036627.1 | 3 - 4 | 486 | 81 | |
XM_006230209.2 | XP_006230271.1 | 5 - 6 | 732 | 81 | |
XM_006230210.2 | XP_006230272.1 | 4 - 5 | 642 | 81 | |
XM_006230211.1 | XP_006230273.1 | 4 - 5 | 497 | 81 | |
GenBank mRNA | BC064440.1 | - | 3 - 4 | 463 | 81 |
CO398643.1 | - | 3 - 4 | 377 | 81 | |
FQ212782.1 | - | 3 - 4 | 437 | 81 | |
FQ213869.1 | - | 3 - 4 | 467 | 81 | |
FQ216820.1 | - | 3 - 4 | 388 | 81 | |
FQ224221.1 | - | 3 - 4 | 477 | 81 | |
FQ228954.1 | - | 3 - 4 | 416 | 81 | |
FQ229100.1 | - | 3 - 4 | 431 | 81 | |
M12919.1 | - | 3 - 4 | 486 | 81 | |
M14420.1 | - | 3 - 4 | 384 | 81 |