The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012963.2 | NP_037095.1 | - | 890 | 70 |
GenBank mRNA | AF275734.1 | - | - | 922 | 70 |
BC061779.1 | - | - | 879 | 70 | |
BC081839.1 | - | - | 890 | 70 | |
BC088402.1 | - | - | 966 | 70 | |
M64986.1 | - | - | 955 | 70 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109373.1 | NP_001102843.1 | 1 - 1 | 934 | 70 |
GenBank mRNA | BC168143.1 | - | 1 - 1 | 970 | 70 |