The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057122.1 | NP_476463.1 | 6 - 7 | 688 | 75 |
GenBank mRNA | BC063145.1 | - | 6 - 7 | 698 | 75 |
D50695.1 | - | 6 - 7 | 688 | 75 | |
FQ225557.1 | - | 6 - 7 | 713 | 75 |