Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ugt2b7
- Entrez Gene ID
-
286989
- Gene Name
- UDP glucuronosyltransferase 2 family, polypeptide B7
- Gene Aliases
-
UDPGT 2B7, Ugt2b8
- Chromosome Location
-
Chr.14: 22597103 - 22619968 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173323.1 |
NP_775445.1 |
2 - 3 |
902 |
65 |
GenBank mRNA |
U27518.1 |
- |
2 - 3 |
902 |
65 |
- Gene Symbol
- LOC689798
- Entrez Gene ID
-
689798
- Gene Name
- similar to UDP-glucuronosyltransferase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 22647498 - 22653977 on Build Rnor_6.0
View Full Product Details