Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kdm5c
Entrez Gene ID
317432
Gene Name
lysine (K)-specific demethylase 5C
Gene Aliases
RGD1560601
Chromosome Location
Chr.X: 22302671 - 22349298 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773146.1 XP_008771368.1 18 - 19 2916 64

View Full Product Details