Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bai3
Entrez Gene ID
301309
Gene Name
brain-specific angiogenesis inhibitor 3
Gene Aliases
-
Chromosome Location
Chr.9: 31280623 - 32022098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106898.1 NP_001100368.1 23 - 24 3401 140
XM_008766928.1 XP_008765150.1 7 - 8 1092 140
XM_008766929.1 XP_008765151.1 8 - 9 985 140
XM_008766930.1 XP_008765152.1 8 - 9 2406 140
XM_008766931.1 XP_008765153.1 8 - 9 1222 140

View Full Product Details