Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dos
Entrez Gene ID
314622
Gene Name
downstream of Stk11
Gene Aliases
-
Chromosome Location
Chr.7: 12433422 - 12441048 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108070.1 NP_001101540.1 7 - 8 1085 59

View Full Product Details