Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shank2
Entrez Gene ID
171093
Gene Name
SH3 and multiple ankyrin repeat domains 2
Gene Aliases
CortBP1, ProSAP1, Spank-3
Chromosome Location
Chr.1: 217173199 - 217593950 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133441.1 NP_597685.1 5 - 6 1108 62
NM_201350.1 NP_958738.1 14 - 15 1957 62
XM_008760068.1 XP_008758290.1 13 - 14 1868 62
GenBank mRNA AF141903.1 - 4 - 5 600 62
AF159048.1 - 4 - 5 618 62
AJ249562.1 - 5 - 6 1108 62
AY298755.1 - 14 - 15 1957 62

View Full Product Details