Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iqsec1
Entrez Gene ID
686590
Gene Name
IQ motif and Sec7 domain 1
Gene Aliases
-
Chromosome Location
Chr.4: 123400730 - 123454656 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006236890.2 XP_006236952.1 11 - 12 3221 67
XM_008763140.1 XP_008761362.1 11 - 12 3221 67
XM_008763141.1 XP_008761363.1 11 - 12 3221 67

View Full Product Details