Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajc8
Entrez Gene ID
313035
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 8
Gene Aliases
-
Chromosome Location
Chr.5: 150725674 - 150743732 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013168.1 NP_001013186.1 4 - 5 319 68
GenBank mRNA BC081863.1 - 4 - 5 319 68
FQ214236.1 - 4 - 5 321 68
FQ231836.1 - 4 - 5 316 68

View Full Product Details