Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxk1
Entrez Gene ID
304298
Gene Name
forkhead box K1
Gene Aliases
RGD1309643
Chromosome Location
Chr.12: 14183436 - 14244634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037219.2 NP_001032296.2 6 - 7 1399 51

View Full Product Details