The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039914.2 | NP_001035003.1 | 4 - 5 | 787 | 76 |
GenBank mRNA | AB234867.1 | - | 4 - 5 | 807 | 76 |