The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024823.4 | NP_001019994.4 | - | 2843 | 81 |
NM_001159653.1 | NP_001153125.1 | - | 1512 | 81 | |
NM_019132.1 | NP_062005.1 | - | 772 | 81 | |
NM_021845.5 | NP_068617.6 | - | 2843 | 81 | |
GenBank mRNA | AF105254.1 | - | - | 1370 | 81 |
AF107844.1 | - | - | 1330 | 81 | |
BC061967.1 | - | - | 734 | 81 | |
BC158589.1 | - | - | 627 | 81 | |
BC168156.1 | - | - | 636 | 81 | |
DQ120475.1 | - | - | 621 | 81 | |
DQ120476.1 | - | - | 621 | 81 | |
FQ214546.1 | - | - | 511 | 81 | |
FQ219804.1 | - | - | 202 | 81 | |
FQ226208.1 | - | - | 300 | 81 | |
M12673.1 | - | - | 836 | 81 | |
M17525.1 | - | - | 772 | 81 |