Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lsm12
Entrez Gene ID
287731
Gene Name
LSM12 homolog (S. cerevisiae)
Gene Aliases
RGD1309441
Chromosome Location
Chr.10: 90105562 - 90127600 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105843.1 NP_001099313.1 4 - 5 538 99

View Full Product Details