The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195616.1 | NP_001182545.1 | 3 - 4 | 1086 | 71 |
XM_006255531.2 | XP_006255593.1 | 3 - 4 | 1321 | 71 | |
XM_006255532.2 | XP_006255594.1 | 3 - 4 | 1229 | 71 | |
XM_006255533.2 | XP_006255595.1 | 3 - 4 | 1165 | 71 | |
GenBank mRNA | BC166813.1 | - | 3 - 4 | 1086 | 71 |