The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014258.1 | NP_001014280.1 | 4 - 5 | 796 | 80 |
XM_006235078.2 | XP_006235140.1 | 4 - 5 | 990 | 80 | |
XM_006235079.2 | XP_006235141.1 | 4 - 5 | 979 | 80 | |
XM_006235080.2 | XP_006235142.1 | 4 - 5 | 479 | 80 | |
GenBank mRNA | BC085849.1 | - | 4 - 5 | 796 | 80 |