Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dennd1b
Entrez Gene ID
289051
Gene Name
DENN/MADD domain containing 1B
Gene Aliases
Fam31b
Chromosome Location
Chr.13: 56015393 - 56242041 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751244.2 XP_003751292.1 13 - 14 1079 64
XM_003751245.2 XP_003751293.1 14 - 15 1139 64
XM_006249933.2 XP_006249995.1 13 - 14 1149 64
XM_006249934.1 XP_006249996.1 14 - 15 1066 64
XM_008769580.1 XP_008767802.1 12 - 13 969 64

View Full Product Details