The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053457.2 | NP_445909.2 | 2 - 3 | 588 | 86 |
GenBank mRNA | AF324043.1 | - | 2 - 3 | 393 | 86 |
BC070927.1 | - | 2 - 3 | 588 | 86 |