The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022289.3 | NP_071625.2 | 7 - 8 | 1227 | 75 |
GenBank mRNA | AF305780.1 | - | 7 - 8 | 1213 | 75 |
BC061554.1 | - | 7 - 8 | 1231 | 75 |