Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm41
Entrez Gene ID
680581
Gene Name
RNA binding motif protein 41
Gene Aliases
-
Chromosome Location
Chr.X: 111272347 - 111325203 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109420.1 NP_001102890.1 5 - 6 986 92

View Full Product Details