Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcaf5
Entrez Gene ID
314273
Gene Name
DDB1 and CUL4 associated factor 5
Gene Aliases
Wdr22
Chromosome Location
Chr.6: 103516586 - 103605270 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100718.1 NP_001094188.1 1 - 2 232 94

View Full Product Details