The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031793.1 | NP_113981.1 | 14 - 15 | 1479 | 138 |
XM_006234324.2 | XP_006234386.1 | 14 - 15 | 1855 | 138 | |
XM_006234325.2 | XP_006234387.1 | 14 - 15 | 1676 | 138 | |
XM_006234326.2 | XP_006234388.1 | 14 - 15 | 1799 | 138 | |
XM_006234327.2 | XP_006234389.1 | 13 - 14 | 1751 | 138 | |
XM_006234328.2 | XP_006234390.1 | 14 - 15 | 1746 | 138 | |
XM_006234330.2 | XP_006234392.1 | 13 - 14 | 1572 | 138 | |
XM_006234331.2 | XP_006234393.1 | 13 - 14 | 1698 | 138 | |
XM_006234333.2 | XP_006234395.1 | 13 - 14 | 1712 | 138 | |
XM_006234334.2 | XP_006234396.1 | 13 - 14 | 1533 | 138 | |
XM_006234335.2 | XP_006234397.1 | 13 - 14 | 1659 | 138 | |
XM_008761930.1 | XP_008760152.1 | 13 - 14 | 1775 | 138 | |
XM_008761931.1 | XP_008760153.1 | 13 - 14 | 1736 | 138 | |
GenBank mRNA | AF043642.1 | - | 14 - 15 | 1479 | 138 |
BC085723.1 | - | 13 - 14 | 1525 | 138 |