Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw10
Entrez Gene ID
303215
Gene Name
F-box and WD repeat domain containing 10
Gene Aliases
-
Chromosome Location
Chr.10: 49259050 - 49296714 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767919.1 XP_008766141.1 9 - 10 2064 71

View Full Product Details