Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt5
Entrez Gene ID
54309
Gene Name
synaptotagmin V
Gene Aliases
SytIX
Chromosome Location
Chr.1: 72860218 - 72867934 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019350.2 NP_062223.1 7 - 8 1143 82
XM_006228287.2 XP_006228349.1 6 - 7 830 82
GenBank mRNA BC092198.1 - 7 - 8 1143 82
U26402.1 - 7 - 8 946 82
X84884.1 - 6 - 7 896 82

View Full Product Details