Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdfy1
Entrez Gene ID
301549
Gene Name
WD repeat and FYVE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 85477070 - 85528890 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008331.1 NP_001008332.1 7 - 8 812 66
XM_006245190.2 XP_006245252.1 8 - 9 896 66
GenBank mRNA BC085807.1 - 7 - 8 812 66

View Full Product Details