The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009674.1 | NP_001009674.1 | 3 - 4 | 526 | 62 |
GenBank mRNA | BC087127.1 | - | 3 - 4 | 526 | 62 |